3-(Hydroxymethyl)indole
Supplier: Molekula
Jump to:
About this item
Synonyms: 3-Indolmethanol, 3-Indolcarbinol, (1H-Indol-3-yl)methanol, Indole-3-methanol, (Indole-3-yl)methanol, 3-Indolemethanol, Indole-3-carbinol
- CAS Number:700-06-1
- Formula:C₉H₉NO
- MDL Number:MFCD00005632
- Molecular Weight:147.18 g/mol
- EINECS:211-836-2
2 Options Available Below
Product Details & Documents
Synonyms: 3-Indolmethanol, 3-Indolcarbinol, (1H-Indol-3-yl)methanol, Indole-3-methanol, (Indole-3-yl)methanol, 3-Indolemethanol, Indole-3-carbinol
- CAS Number:700-06-1
- Formula:C₉H₉NO
- MDL Number:MFCD00005632
- Molecular Weight:147.18 g/mol
- EINECS:211-836-2



