

(2,4-Dichlorophenoxy)acetic acid, tan powder
Catalog # ICNA0210155380
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:221.04 g/mol
Merck Index:13,02825
Boiling Pt:346 °C (1013 hPa)
Melting Pt:134…140 °C
Density:1,563 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-361-1
UN:3077
ADR:6.1,III
:GHS05
:GHS07
CAS Number:94-75-7
Formula:(C₆H₃Cl₂)OCH₂COOH
MDL Number:MFCD00004300