Nitric acid ≥70%
Glass bottle, 2,5 L
Catalog # HONE17512.2500
Supplier: Honeywell Chemicals
Jump to:
Product Details & Documents
Synonyms: Nitric acid solution
- CAS Number:7697-37-2
- Formula:HNO₃
- MDL Number:MFCD00011349
- Molecular Weight:63.01 g/mol
- Storage Temperature:Ambient
- ADR:8,I
- :Density: 1.51 g/cm³ (20 °C)
- EINECS:231-714-2
- UN:2031
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



