1,3-Dimethylurea
Catalog # EHERC12760000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:88.11 g/mol
Boiling Pt:268 °C (1013 hPa)
Melting Pt:96 °C
Density:1.142 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-498-7
CAS Number:96-31-1
Formula:C₃H₈N₂O
MDL Number:MFCD00008286