Order Entry
Export
ContactUsLinkComponent
4-Nitro-o-xylene
Irritant
4-Nitro-o-xylene
Catalog # EHERC12727920
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
4-Nitro-o-xylene
Catalog # EHERC12727920
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :151.16 g/mol
Boiling Pt:254 °C (1013 hPa)
Melting Pt:31…32 °C
Density:1.117 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-761-6
UN:3447
ADR:6.1,II
 :GHS07
CAS Number:99-51-4
Formula:C₈H₉NO₂
MDL Number:MFCD00007268