2,6-Diisopropylnaphthalene
Catalog # EHERC12637700
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:212.33 g/mol
Boiling Pt:279.3 °C (1013 hPa)
Melting Pt:67…69 °C
Density:0.95 g/cm³ (20 °C)
EINECS:246-045-1
CAS Number:24157-81-1
Formula:C₁₆H₂₀
MDL Number:MFCD00021648