4,4'-Dibromobenzophenone
Catalog # EHERC12220000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:340.01 g/mol
EINECS:223-632-0
CAS Number:3988-03-2
Formula:C₁₃H₈Br₂O
MDL Number:MFCD00016330