3-Methoxypropyl isothiocyanate
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:17702-11-3
:
Formula:C₅H₉NOS
MDL Number:MFCD00022063
Boiling Pt:86…90 °C (22 mmHg)
Density:1.0408 g/cm³ (20 °C)
UN:2810
ADR:6.1,III
:131.2 g/mol