Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Fluoro-o-toluonitrile
5-Fluoro-o-toluonitrile
5-Fluoro-o-toluonitrile
5-Fluoro-o-toluonitrile
Supplier:
Thermo Scientific
Certificates
Expand image
5-Fluoro-o-toluonitrile
Supplier:
Thermo Scientific
About this item
CAS Number:
77532-79-7
:
Formula:
C₈H₆FN
MDL Number:
MFCD00042295
:
135.14 g/mol
Boiling Pt:
226 °C (1013 hPa)
Melting Pt:
42…46 °C
UN:
3439
ADR:
6.1,III