Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Bis(trifluoromethyl)benzylamine
3,5-Bis(trifluoromethyl)benzylamine
3,5-Bis(trifluoromethyl)benzylamine
3,5-Bis(trifluoromethyl)benzylamine
Supplier:
Thermo Scientific
Certificates
Expand image
3,5-Bis(trifluoromethyl)benzylamine
Supplier:
Thermo Scientific
About this item
CAS Number:
85068-29-7
:
Formula:
C₉H₇F₆N
MDL Number:
MFCD00009909
Boiling Pt:
82…84 °C (15 mmHg)
Melting Pt:
46…55 °C
:
243.15 g/mol
EINECS:
285-290-9