3,5-Bis(trifluoromethyl)benzaldehyde
Catalog # MAYBSEW00692.1
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:242.12 g/mol
Boiling Pt:47…48 °C (0.2 mmHg)
Boiling Pt:47…48 °C (0,2 mmHg)
Melting Pt:–2 °C
Density:1,46 g/cm³ (20 °C)
CAS Number:401-95-6
Formula:C₉H₄F₆O
MDL Number:MFCD00010206