Order Entry
Export
Orders LinkContactUsLinkComponent
Methyl propiolate

Methyl propiolate

Supplier:   Thermo Scientific
Methyl propiolate
Supplier:  Thermo Scientific
Certificates
Methyl propiolate

3 Options Available Below

Product Details & Documents
Synonyms:  Methyl acetylenecarboxylate, Propiolic acid methyl ester, 2-Propynoic acid methyl ester
  • CAS Number:922-67-8
  • Formula:C₄H₄O₂
  • MDL Number:MFCD00008572
  • Molecular Weight:84.07 g/mol
  • Boiling Pt:105 °C (1013 hPa)
  • Density:1.196 g/cm³ (20 °C)
  • EINECS:213-083-5
  • UN:3272
  • ADR:3,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g