Order Entry
Export
Orders LinkContactUsLinkComponent
 

3,4,5-Trimethoxybenzyl alcohol

Supplier:   Thermo Scientific
3,4,5-Trimethoxybenzyl alcohol
Supplier:  Thermo Scientific
Certificates
3,4,5-Trimethoxybenzyl alcohol

3 Options Available Below

Product Details & Documents
Synonyms:  2,4,6-Trimethoxybenzyl alcohol, (3,4,5-Trimethoxyphenyl)methanol
  • CAS Number:3840-31-1
  • Formula:C₁₀H₁₄O₄
  • MDL Number:MFCD00060356
  • Molecular Weight:198.22 g/mol
  • Boiling Pt:350 °C (1013 hPa)
  • Melting Pt:36…38 °C
  • Density:1.1427 g/cm³ (20 °C)
  • EINECS:223-331-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g