

α,3,4-Trichlorotoluene
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:102-47-6
:
Formula:C₇H₅Cl₃
MDL Number:MFCD00000906
:195.47 g/mol
Boiling Pt:241 °C (1013 hPa)
Melting Pt:37.5 °C
Density:1.41 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-033-0
UN:3265
ADR:8,II
:GHS05
:GHS07