
α,2,6-Trichlorotoluene
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:2014-83-7
:
Formula:C₇H₅Cl₃
MDL Number:MFCD00000897
:195.47 g/mol
Boiling Pt:248 °C (1013 hPa)
Melting Pt:49…50 °C
EINECS:217-940-4
UN:3261
ADR:8,II
:GHS05