3'-(Trifluoromethyl)acetophenone
Supplier: Thermo Scientific
Jump to:
About this item
Synonyms: m-(Trifluoromethyl)acetophenone, 1-[3-(Trifluoromethyl)phenyl]ethan-1-one
- CAS Number:349-76-8
- Formula:C₉H₇F₃O
- MDL Number:MFCD00000391
- Molecular Weight:188.15 g/mol
- Boiling Pt:198 °C (1013 hPa)
- Density:1,23 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:206-490-4
3 Options Available Below
Product Details & Documents
Synonyms: m-(Trifluoromethyl)acetophenone, 1-[3-(Trifluoromethyl)phenyl]ethan-1-one
- CAS Number:349-76-8
- Formula:C₉H₇F₃O
- MDL Number:MFCD00000391
- Molecular Weight:188.15 g/mol
- Boiling Pt:198 °C (1013 hPa)
- Density:1,23 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:206-490-4
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



