Order Entry
Export
Orders LinkContactUsLinkComponent
4-Iodopyridine

4-Iodopyridine

Supplier:   CHEMPUR
4-Iodopyridine
Supplier:  CHEMPUR
Certificates

2 Options Available Below

Product Details & Documents
  • CAS Number:15854-87-2
  • Formula:C₅H₄IN
  • MDL Number:MFCD02093937
  • Molecular Weight:205 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

This product has been blocked by your organization. Please contact your purchasing department for more information