Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3,5-dihydroxybenzoate
Methyl-3,5-dihydroxybenzoate
Methyl-3,5-dihydroxybenzoate
Methyl-3,5-dihydroxybenzoate
Supplier:
CHEMPUR
Certificates
Expand image
Methyl-3,5-dihydroxybenzoate
Supplier:
CHEMPUR
About this item
CAS Number:
2150-44-9
:
Formula:
C₈H₈O₄
MDL Number:
MFCD00002289
:
168.15 g/mol
Boiling Pt:
300 °C (1013 hPa)
Melting Pt:
168…170 °C
EINECS:
218-426-2