ω-Bromo-4-methylacetophenone
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:619-41-0
:
Formula:C₉H₉BrO
MDL Number:MFCD00000203
:213.07 g/mol
Boiling Pt:105 °C (0,1 mmHg)
Melting Pt:95…97 °C
Storage Temperature:Refrigerator
EINECS:210-595-0