Order Entry
Export
Orders LinkContactUsLinkComponent
2-Thiophenemethanol

2-Thiophenemethanol

2-Thiophenemethanol
Certificates
2-Thiophenemethanol

3 Options Available Below

 
Synonyms:  Thiophen-2-methanol, Thiophen-2-ylmethanol, 2-Thienylmethanol
  • CAS Number:636-72-6
  • Formula:C₅H₆OS
  • MDL Number:MFCD00005454
  • Molecular Weight:114.17 g/mol
  • Boiling Pt:207 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1.206 g/cm³ (20 °C)
  • EINECS:211-264-3
Product Family Options

Product Information

  • Pack type
    Pk
     
     
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g