Order Entry
Export
Orders LinkContactUsLinkComponent
ω-Bromo-4-fluoroacetophenone

ω-Bromo-4-fluoroacetophenone

Supplier:   Thermo Scientific
ω-Bromo-4-fluoroacetophenone
Supplier:  Thermo Scientific
Certificates
ω-Bromo-4-fluoroacetophenone

3 Options Available Below

Product Details & Documents
Synonyms:  Bromomethyl p-fluorophenyl ketone, α-Bromo-4-fluoroacetophenone, 2-Bromo-1-(4-fluorophenyl)ethan-1-one, 2-Bromo-4'-fluoroacetophenone, 4-Fluorophenacyl bromide, 2-Bromo-1-(4-fluorophenyl)ethanone
  • CAS Number:403-29-2
  • Formula:C₈H₆BrFO
  • MDL Number:MFCD00040830
  • Molecular Weight:217.04 g/mol
  • Boiling Pt:82 °C (0,4 mmHg)
  • Boiling Pt:82 °C (0.4 mmHg)
  • Melting Pt:49 °C
  • EINECS:206-955-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g