

3,5-Bis(trifluoromethyl)pyridine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:20857-47-0
:
Formula:C₇H₃F₆N
MDL Number:MFCD00128904
Boiling Pt:117…118 °C
Melting Pt:35…38 °C
Flash Pt:>100 °C (212 °F)
UN:2810
ADR:6.1,II
:215.1 g/mol
:GHS06
:GHS07
Storage Temperature:Ambient