0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2,5-Dibromobenzaldehyde
2,5-Dibromobenzaldehyde ≥97%
2,5-Dibromobenzaldehyde ≥97%
Irritant
2,5-Dibromobenzaldehyde ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
2,5-Dibromobenzaldehyde ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
74553-29-0
:
Formula:
C₇H₄Br₂O
MDL Number:
MFCD00462864
:
263.92 g/mol
Melting Pt:
88…102 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH31689
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation