
2,4-Dimethoxybenzylamine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:20781-20-8
:
Formula:C₉H₁₃NO₂
MDL Number:MFCD00052393
:167.21
Boiling Pt:140 °C (1 mmHg)
Density:1.113
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
ADR:8,III
:GHS05