Order Entry
Export
ContactUsLinkComponent
1,4-Difluoro-2-iodobenzene ≥97%
Irritant
1,4-Difluoro-2-iodobenzene ≥97%
Catalog # H26890.06
CAS Number:  
Irritant
1,4-Difluoro-2-iodobenzene ≥97%
Catalog # H26890.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

CAS Number:2265-92-1
 :
Formula:C₆H₃F₂I
MDL Number:MFCD03094151
 :239.99 g/mol
Boiling Pt:178…179 °C
Density:1.998
Flash Pt:85 °C (185 °F)
 :GHS07
Storage Temperature:Ambient