
3,4-Bis(trifluoromethyl)pyridine 96%
Catalog # H26008.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
CAS Number:20857-46-9
:
Formula:C₇H₃F₆N
MDL Number:MFCD08686956
:215.1 g/mol
Boiling Pt:42…44 °C (24 mmHg)
:GHS07
Storage Temperature:Ambient