Order Entry
Export
ContactUsLinkComponent
4,4-Dimethylcyclohexanone ≥98%
4,4-Dimethylcyclohexanone ≥98%
Catalog # H25857.03
CAS Number:  
4,4-Dimethylcyclohexanone ≥98%
Catalog # H25857.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

CAS Number:4255-62-3
 :
Formula:C₈H₁₄O
MDL Number:MFCD00234965
 :126.2 g/mol
Melting Pt:41…43 °C
Flash Pt:62 °C (144 °F)
Storage Temperature:Ambient