
α,α,α',α'-Tetrafluoro-p-xylene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:369-54-0
:
Formula:C₈H₆F₄
MDL Number:MFCD01320708
:178.13 g/mol
Boiling Pt:83…84 °C (30 mmHg)
Density:1.312
Flash Pt:63 °C (145 °F)
:GHS07
Storage Temperature:Ambient