Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dichlorosalicylaldehyde ≥98%

3,5-Dichlorosalicylaldehyde ≥98%

3,5-Dichlorosalicylaldehyde ≥98%
Certificates
3,5-Dichlorosalicylaldehyde ≥98%
3,5-Dichlorosalicylaldehyde ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 90-60-8 Molecular Formula: C7H4Cl2O2 Formula Weight: 191.01 Melting Point: 94-97° Beilstein Reference: 973391

Synonyms:  3,5-Dichloro-2-hydroxybenzaldehyde
  • CAS Number:90-60-8
  • Formula:C₇H₄Cl₂O₂
  • MDL Number:MFCD00003320
  • Molecular Weight:191.01 g/mol
  • Boiling Pt:723 °C (1013 hPa)
  • Melting Pt:94…97 °C
  • Storage Temperature:Ambient
  • EINECS:202-005-5
Product Family Options

Product Information