0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1,3-Diphenyl-2-thiourea
1,3-Diphenyl-2-thiourea ≥98%
1,3-Diphenyl-2-thiourea ≥98%
:
Thermo Fisher Scientific
1,3-Diphenyl-2-thiourea ≥98%
:
Thermo Fisher Scientific
:
102-08-9
:
:
C₁₃H₁₂N₂S
:
MFCD00004921
:
228.32 g/mol
:
14,3333
:
365 °C (1013 hPa)
:
150…154 °C
:
1.320
:
Ambient
:
203-004-2
:
2811
:
6.1,II
:
GHS06
:
GHS07
msds_ALFAA19644
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years