

α,α,α,α',α',α'-Hexafluoro-p-xylene ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:433-19-2
:
Formula:C₈H₄F₆
MDL Number:MFCD00000402
:214.11 g/mol
Boiling Pt:116…117 °C
Melting Pt:–1 °C
Density:1.381
Flash Pt:21 °C (69 °F)
Storage Temperature:Ambient
EINECS:207-086-0
ADR:3,II
:GHS02
:GHS07