
Oleyl alcohol tech. 80-85%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:143-28-2
:
Formula:C₁₈H₃₆O
MDL Number:MFCD00002993
:268.48 g/mol
Merck Index:14,6831
Boiling Pt:206…207 °C (13 mmHg)
Melting Pt:∼ 6 °C
Density:0.849
Flash Pt:180 °C (356 °F)
EINECS:205-597-3
:GHS07
Storage Temperature:Ambient