0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Tris(3-methoxyphenyl)phosphine
Tris(3-methoxyphenyl)phosphine ≥98%
Tris(3-methoxyphenyl)phosphine ≥98%
Tris(3-methoxyphenyl)phosphine ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Tris(3-methoxyphenyl)phosphine ≥98%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
29949-84-6
:
Formula:
C₂₁H₂₁O₃P
MDL Number:
MFCD00008386
:
352.37 g/mol
Melting Pt:
113…115 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFAA17949
Videos
Thermo Scientific Acroseal 25 Years
Documentation