
3,5-Bis(trifluoromethyl)benzonitrile ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:27126-93-8
:
Formula:C₉H₃F₆N
MDL Number:MFCD00000379
:239.12 g/mol
Boiling Pt:154…156 °C
Melting Pt:16…20 °C
Density:1.421
Flash Pt:72 °C (161 °F)
Storage Temperature:Ambient
EINECS:248-240-7
UN:3276
ADR:6.1,III
:GHS07