Order Entry
Export
Orders LinkContactUsLinkComponent
Methyl propiolate ≥99%

Methyl propiolate ≥99%

Flammable
Irritant
Methyl propiolate ≥99%
Certificates
Methyl propiolate ≥99%
Methyl propiolate ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 922-67-8 Molecular Formula: HC≡CCO2CH3 Formula Weight: 84.07 Flash Point: 10°(50°F) Refractive Index: 1.4080 Odour: Pungent Boiling Point: 98-101° Beilstein Reference: 605462 UN No.: 3272 MDL No.: MFCD00008572

Synonyms:  Methyl acetylenecarboxylate, Propiolic acid methyl ester, 2-Propynoic acid methyl ester
  • CAS Number:922-67-8
  • Formula:C₄H₄O₂
  • MDL Number:MFCD00008572
  • Molecular Weight:84.07 g/mol
  • Boiling Pt:98…101 °C
  • Density:1.016
  • Flash Pt:10 °C (50 °F)
  • Storage Temperature:Ambient
  • EINECS:213-083-5
  • UN:3272
  • ADR:3,II
Product Family Options

Product Information