3-Methylpyrocatechol ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 488-17-5 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 140°(284°F) Melting Point: 62-68° Boiling Point: 241° Beilstein Reference: 774602 MDL No.: MFCD00016435
Synonyms: 3-Methylcatechol, 3-Methylbenzene-1,2-diol
- CAS Number:488-17-5
- Formula:C₇H₈O₂
- MDL Number:MFCD00016435
- Molecular Weight:124.14 g/mol
- Boiling Pt:241 °C
- Melting Pt:62…68 °C
- Flash Pt:140 °C (284 °F)
- Storage Temperature:Ambient
- EINECS:207-672-6
2 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 488-17-5 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 140°(284°F) Melting Point: 62-68° Boiling Point: 241° Beilstein Reference: 774602 MDL No.: MFCD00016435
Synonyms: 3-Methylcatechol, 3-Methylbenzene-1,2-diol
- CAS Number:488-17-5
- Formula:C₇H₈O₂
- MDL Number:MFCD00016435
- Molecular Weight:124.14 g/mol
- Boiling Pt:241 °C
- Melting Pt:62…68 °C
- Flash Pt:140 °C (284 °F)
- Storage Temperature:Ambient
- EINECS:207-672-6



