Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Ethylpyrocatechol ≥98%

4-Ethylpyrocatechol ≥98%
Certificates
4-Ethylpyrocatechol ≥98%
4-Ethylpyrocatechol ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 1124-39-6 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 37-40° Boiling Point: 105-107°/0.5mm MDL No.: MFCD00015847

Synonyms:  4-Ethylbenzene-1,2-diol, 4-Ethylcatechol
  • CAS Number:1124-39-6
  • Formula:C₈H₁₀O₂
  • MDL Number:MFCD00015847
  • Molecular Weight:138.17 g/mol
  • Boiling Pt:105…107 °C (0.5 mmHg)
  • Melting Pt:37…40 °C
  • Storage Temperature:Ambient
  • EINECS:214-397-5
Product Family Options

Product Information