Rhodium ≥99.8% (metals basis), foil, Thickness: 0.025 mm (0.001 in)
Catalog # 45640.FF
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:1643
- Pk Info:25×25 mm
Specifications
About this item
:
Merck Index:12,08353
Boiling Pt:3400…3700 °C (1013 hPa)
Melting Pt:1960 °C
Density:12.41 g/cm³ (20 °C)
:102.9 g/mol
Storage Temperature:Ambient
EINECS:231-125-0
CAS Number:7440-16-6
Formula:Rh
MDL Number:MFCD00011201