Order Entry
Export
ContactUsLinkComponent
2,6-Dimethyl-5-heptenal stabilised
2,6-Dimethyl-5-heptenal stabilised
Catalog # 17773.30
CAS Number:  
2,6-Dimethyl-5-heptenal stabilised
Catalog # 17773.30
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

About this item

 :
 :140.23 g/mol
Boiling Pt:116…124 °C (100 mmHg)
Density:0.851
Flash Pt:63 °C (145 °F)
Storage Temperature:Ambient
EINECS:203-427-2
UN:1989
ADR:,
CAS Number:106-72-9
Formula:C₉H₁₆O
MDL Number:MFCD00006981