Zinc iodide ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Synonyms: Zinc (II) iodide
- CAS Number:10139-47-6
- Formula:I₂Zn
- MDL Number:MFCD00011299
- Molecular Weight:319.2 g/mol
- Merck Index:14,10140
- Boiling Pt:625 °C (dec.)
- Melting Pt:446 °C
- Density:4.74
- Storage Temperature:Ambient
- EINECS:233-396-0
- UN:3260
- ADR:8,II
2 Options Available Below
Product Details & Documents
Synonyms: Zinc (II) iodide
- CAS Number:10139-47-6
- Formula:I₂Zn
- MDL Number:MFCD00011299
- Molecular Weight:319.2 g/mol
- Merck Index:14,10140
- Boiling Pt:625 °C (dec.)
- Melting Pt:446 °C
- Density:4.74
- Storage Temperature:Ambient
- EINECS:233-396-0
- UN:3260
- ADR:8,II



