3,5-Dichlorosalicylaldehyde 98+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 3,5-Dichloro-2-hydroxybenzaldehyde
- CAS Number:90-60-8
- Formula:C₇H₄Cl₂O₂
- MDL Number:MFCD00003320
- Molecular Weight:191.01 g/mol
- Boiling Pt:723 °C (1013 hPa)
- Melting Pt:91…94 °C
- Storage Temperature:Ambient
- EINECS:202-005-5
2 Options Available Below
Product Details & Documents
Synonyms: 3,5-Dichloro-2-hydroxybenzaldehyde
- CAS Number:90-60-8
- Formula:C₇H₄Cl₂O₂
- MDL Number:MFCD00003320
- Molecular Weight:191.01 g/mol
- Boiling Pt:723 °C (1013 hPa)
- Melting Pt:91…94 °C
- Storage Temperature:Ambient
- EINECS:202-005-5
Documents
ACRO40700
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:91 to 94 °C
- Titration with TBAH:≥98.0 %



