Oil of castor
: Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi
:8001-79-4
:
:MFCD00130746
:229 °C (760 mmHg)
:–10 °C
:0.957 g/cm³
:229 °C
:13,01908
:Ambient
:232-293-8
- Environmentally Preferable:
- Appearance:Clear colorless to yellow Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4770 to 1.4810 (20°C, 589 nm)
- Acid value:≤2 mg KOH/g
- Hydroxyl value:≥150 mg KOH/g
- Iodine value:82 to 90 g I2/100g