
5-Bromo-2,4-dichloropyrimidine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:36082-50-5
:
Formula:C₄HBrCl₂N₂
MDL Number:MFCD00127818
:227.87 g/mol
Boiling Pt:128 °C (15 mmHg)
Melting Pt:29…30 °C
Density:1.781 g/cm³
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Clear to slightly hazy colorless to light yellow Liquid or low melting solid
- Infrared spectrum:Conforms
- Refractive index:1.6020 to 1.6050 (20°C, 589 nm)
- GC:≥97.5 %