Toluidine blue O, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:92-31-9
:
Formula:C₁₅H₁₆ClN₃S
MDL Number:MFCD00011934
:305.83 g/mol
Merck Index:12,09658
Storage Temperature:Ambient
EINECS:202-146-2
Specification Test Results
- Appearance:Dark green Powder
- Infrared spectrum:Conforms
- UV spectrum:Complies