2-Mercaptobenzothiazole 97%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 2-Benzothiazolethiol, Mercapto-2 benzothiazole, Benzo[d]thiazole-2(3H)-thione, Benzothiazole-2-thiol, MBT
- CAS Number:149-30-4
- Formula:C₇H₅NS₂
- MDL Number:MFCD00005781
- Molecular Weight:167.26 g/mol
- Merck Index:13,05893
- Boiling Pt:302 °C (1013 hPa)
- Melting Pt:172…182 °C
- Density:1,42 g/cm³ (20 °C)
- Flash Pt:243 °C
- Storage Temperature:Ambient
- EINECS:205-736-8
- UN:3077
- ADR:9,III
4 Options Available Below
Product Details & Documents
Synonyms: 2-Benzothiazolethiol, Mercapto-2 benzothiazole, Benzo[d]thiazole-2(3H)-thione, Benzothiazole-2-thiol, MBT
- CAS Number:149-30-4
- Formula:C₇H₅NS₂
- MDL Number:MFCD00005781
- Molecular Weight:167.26 g/mol
- Merck Index:13,05893
- Boiling Pt:302 °C (1013 hPa)
- Melting Pt:172…182 °C
- Density:1,42 g/cm³ (20 °C)
- Flash Pt:243 °C
- Storage Temperature:Ambient
- EINECS:205-736-8
- UN:3077
- ADR:9,III
Documents
ACRO33205
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- Melting point:177 to 182 °C
- Solubility:(2.5% in DMSO) Clear colourless to yellow solution
- Titration with NaOH:96.0 to 104.0 %



