
Ethyl-3,4-dihydroxybenzoate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3943-89-3
:
Formula:C₉H₁₀O₄
MDL Number:MFCD00002199
:182.18 g/mol
Melting Pt:131…136 °C
EINECS:223-529-0
:GHS07
Specification Test Results
- Appearance:Light yellow to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:131 to 136 °C
- GC:≥96.0 %