

N-Methyl-N-(trimethylsilyl)trifluoroacetamide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:24589-78-4
:
Formula:C₆H₁₂F₃NOSi
MDL Number:MFCD00000411
:199.25 g/mol
Boiling Pt:130…132 °C (760 mmHg)
Density:1.07 g/cm³
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:246-331-6
UN:1993
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %