ContactUsLinkComponent
Benzyltrimethylammonium chloride 98+%
Toxic
Health
Benzyltrimethylammonium chloride 98+%
Certificates
undefined
Toxic
Health
Benzyltrimethylammonium chloride 98+%
Specification Test Results
  • Appearance:
    White to cream Crystalline powder and/or chunks
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥98.0 %