

2-(Trimethylsilyl)ethanol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2916-68-9
:
Formula:HOH₂CCH₂Si(CH₃)₃
MDL Number:MFCD00002825
:118.25 g/mol
Boiling Pt:71…73 °C (35 mmHg)
Melting Pt:0 °C
Density:0.825 g/cm³
Flash Pt:50 °C
Storage Temperature:Ambient
EINECS:220-844-5
UN:1987
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4235 to 1.4255 (20°C, 589 nm)
- GC:≥98.5 %