Order Entry
Export
Orders LinkContactUsLinkComponent
Methyl propiolate 99%

Methyl propiolate 99%

Irritant
Flammable
Methyl propiolate 99%
Certificates
Methyl propiolate 99%

3 Options Available Below

Product Details & Documents
Synonyms:  Methyl acetylenecarboxylate, Propiolic acid methyl ester, 2-Propynoic acid methyl ester
  • CAS Number:922-67-8
  • Formula:C₄H₄O₂
  • MDL Number:MFCD00008572
  • Molecular Weight:84.07 g/mol
  • Boiling Pt:105 °C (1013 hPa)
  • Density:0.94 g/cm³
  • Flash Pt:10 °C
  • Storage Temperature:Refrigerator
  • EINECS:213-083-5
  • UN:3272
  • ADR:3,II

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4070 to 1.4090 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Chloride (Cl):
    8.6 to 9.6 %

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 99,40 €
    99,40 €
    1 * 10 g
       99,40 €

    Specifications:
    More
    Specifications
    Cat. No.
    ACRO174040100
  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 321,00 €
    321,00 €
    1 * 50 g
       321,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ACRO174040500
  • Glass bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 1 020,00 €
    1 020,00 €
    1 * 250 g
       1 020,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ACRO174042500